missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyltrimethylammonium Chloride 95.0+%, TCI America™

Catalog No. p-7130324
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H008225G 25 g
H0082100G 100 g
H0082500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H008225G Supplier TCI America Supplier No. H008225G
Only null left

Chemical Identifiers

CAS 112-02-7
Molecular Formula C19H42ClN
Molecular Weight (g/mol) 320.00
MDL Number MFCD00011773
InChI Key WOWHHFRSBJGXCM-UHFFFAOYSA-M
Synonym hexadecyltrimethylammonium chloride, cetrimonium chloride, cetyltrimethylammonium chloride, n,n,n-trimethylhexadecan-1-aminium chloride, dehyquart a, cetyl trimethyl ammonium chloride, aliquat 6, trimethylcetylammonium chloride, arquad 16-50, n-hexadecyltrimethylammonium chloride
PubChem CID 8154
ChEBI CHEBI:53581
IUPAC Name hexadecyltrimethylazanium chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C

Specifications

Melting Point 234°C
Color White
pH 4.0 to 7.5 (50g/L H2O 25°C)
Quantity 25 g
UN Number 3077
Formula Weight 320.00
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyltrimethylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber ML9145000
TSCA Yes