missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecyl Palmitate 97.0+%, TCI America™
$106.33 - $814.51
Chemical Identifiers
| CAS | 540-10-3 |
|---|---|
| Molecular Formula | C32H64O2 |
| Molecular Weight (g/mol) | 480.86 |
| MDL Number | MFCD00053739 |
| InChI Key | PXDJXZJSCPSGGI-UHFFFAOYSA-N |
| Synonym | cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester |
| PubChem CID | 10889 |
| ChEBI | CHEBI:75584 |
| IUPAC Name | hexadecyl hexadecanoate |
| SMILES | CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC |
Chemical Identifiers
| C32H64O2 | |
| MFCD00053739 | |
| cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester | |
| CHEBI:75584 | |
| CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC |
Specifications
| 540-10-3 | |
| White | |
| C32H64O2 | |
| cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester | |
| CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC | |
| 480.86 | |
| CHEBI:75584 | |
| ≥97.0% (GC) | |
| Hexadecyl Palmitate |
| 53°C | |
| 534°C | |
| MFCD00053739 | |
| PXDJXZJSCPSGGI-UHFFFAOYSA-N | |
| hexadecyl hexadecanoate | |
| 10889 | |
| 480.86 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2489
RTECSNumber : RT4750000
TSCA : Yes