missing translation for 'onlineSavingsMsg'
Learn More

Hexadecyl Palmitate 97.0+%, TCI America™

Catalog No. P107725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P107725G 25 g
P1077500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P107725G Supplier TCI America Supplier No. P107725G

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32H64O2
Molecular Weight (g/mol) 480.86
MDL Number MFCD00053739
InChI Key PXDJXZJSCPSGGI-UHFFFAOYSA-N
Synonym cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PubChem CID 10889
ChEBI CHEBI:75584
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 53°C
Color White
Boiling Point 534°C
Quantity 25 g
Formula Weight 480.86
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyl Palmitate
EINECSNumber (2)-2489
RTECSNumber RT4750000
TSCA Yes