missing translation for 'onlineSavingsMsg'
Learn More

Hexabromobenzene 99.0+%, TCI America™

Catalog No. p-7130510
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H036325G 25 g
H0363500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H036325G Supplier TCI America Supplier No. H036325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-82-1
Molecular Formula C6Br6
Molecular Weight (g/mol) 551.49
MDL Number MFCD00000058
InChI Key CAYGQBVSOZLICD-UHFFFAOYSA-N
Synonym hexabromobenzene, benzene, hexabromo, perbromobenzene, unii-t01859xwir, ccris 5917, benzene, 1,2,3,4,5,6-hexabromo, hxbbz, hexabioomobenzene, acmc-20aj0o
PubChem CID 6905
IUPAC Name 1,2,3,4,5,6-hexabromobenzene
SMILES C1(=C(C(=C(C(=C1Br)Br)Br)Br)Br)Br

Specifications

Quantity 25 g
Formula Weight 551.49
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexabromobenzene
EINECSNumber (3)-0059
TSCA Yes