missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, Dodecahydrate, Crystal, ACS, 98.5 -102.0%, Spectrum™ Chemical

Catalog No. 18610474
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610474 50 kg Poly Bottle
18610469 25 g Poly Bottle
18610470 125 g Poly Bottle
18610471 500 g Poly Bottle
18610472 2.5 kg Poly Bottle
18610473 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610474 Supplier Spectrum Chemical Mfg Cor Supplier No. F100550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.18
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
IUPAC Name iron(3+) ammonium dodecahydrate disulfate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Violet
CAS Max % 1
Percent Purity 98.5 to 102.0%
Grade ACS Reagent
Packaging Poly Bottle
Quantity 50 kg
Chemical Name or Material Ferric Ammonium Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.