missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HERC4 Monoclonal Antibody (K29058_14H6), Invitrogen™
Mouse Monoclonal Antibody
$1,421.98 - $2,134.69
Specifications
| Antigen | HERC4 |
|---|---|
| Clone | K29058_14H6 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: IVLEPRKSDF FINKRVRDVG CGLRHTVFVL DDGTVYTCGC NDLGQLGHEK SRKKPEQVVA LDAQNIVAVS CGEAHTLALN DKGQVYAWGL DSDGQLGLVG SEECIRVPRN IKSLSDIQIV QVACGYYHSL ALSKASEVFC WGQNKYGQLG LGTDCKKQTS PQLLKSLLGI PFMQVAAGGA HSFVLTLSGA IFGWGRNKFG QLGLNDENDR YVPNLLKSLR SQKIVYICCG EDHTAALTKE
HERC4 Monoclonal antibody specifically detects HERC4 in Human, Rat samples. It is validated for Western BlotSpecifications
| HERC4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide | |
| 1700056O17Rik; 4921531D01Rik; 9530080M15Rik; HECT and RLD domain containing E3 ubiquitin protein ligase 4; HECT domain and RCC1-like domain-containing protein 4; hect domain and RLD 4; HECT-type E3 ubiquitin transferase HERC4; HERC4; KIAA1593; mKIAA1593; Probable E3 ubiquitin-protein ligase HERC4 | |
| Recombinant human HERC4 (immunogen range is 21-260). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Herc4 |
| K29058_14H6 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| Q5GLZ8, Q5PQN1 | |
| 26091, 309758 | |
| IgG1 | |
| Protein A | |
| Antibody |