missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HERC4 Monoclonal Antibody (K29058_14H6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556026
Description
Sequence of this protein is as follows: IVLEPRKSDF FINKRVRDVG CGLRHTVFVL DDGTVYTCGC NDLGQLGHEK SRKKPEQVVA LDAQNIVAVS CGEAHTLALN DKGQVYAWGL DSDGQLGLVG SEECIRVPRN IKSLSDIQIV QVACGYYHSL ALSKASEVFC WGQNKYGQLG LGTDCKKQTS PQLLKSLLGI PFMQVAAGGA HSFVLTLSGA IFGWGRNKFG QLGLNDENDR YVPNLLKSLR SQKIVYICCG EDHTAALTKE
Probable E3 ubiquitin-protein ligase involved in either protein trafficking or in the distribution of cellular structures. Required for spermatozoon maturation and fertility, and for the removal of the cytoplasmic droplet of the spermatozoon. E3 ubiquitin-protein ligases accept ubiquitin from an E2 ubiquitin- conjugating enzyme in the form of a thioester and then directly transfer it to targeted substrates.Specifications
| HERC4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q5GLZ8, Q5PQN1 | |
| Herc4 | |
| Recombinant human HERC4 (immunogen range is 21-260). | |
| 200 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K29058_14H6 | |
| Unconjugated | |
| Herc4 | |
| 1700056O17Rik; 4921531D01Rik; 9530080M15Rik; HECT and RLD domain containing E3 ubiquitin protein ligase 4; HECT domain and RCC1-like domain-containing protein 4; hect domain and RLD 4; HECT-type E3 ubiquitin transferase HERC4; HERC4; KIAA1593; mKIAA1593; Probable E3 ubiquitin-protein ligase HERC4 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26091, 309758 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |