missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Honeywell Riedel-de Haën™
Puriss. p.a., Reag. Ph. Eur.,≥99% n-heptane basis (GC)
Supplier: Honeywell Chemicals 3228725L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Heptane | |
| Reag. Ph. Eur. | |
| Colorless liquid | |
| 98.2°C to 98.4°C | |
| 0.07 | |
| 433°F | |
| n20/D 1.3870 to 1.3880 | |
| Free acid (as CH3COOH): ≤0.0005% | |
| CH3(CH2)5CH3 | |
| 40mmHg (20°C), 83mmHg (37.7°C) | |
| 1730763 | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| heptane | |
| 8900 | |
| 100.2g/mol | |
| puriss. p.a. |
| -91°C | |
| 142-82-5 | |
| 0.684g/mL at 25°C | |
| 2.5 L | |
| −4°C (24.8°F) | |
| Glass Bottle | |
| 0 to 0.001% | |
| C7H16 | |
| MFCD00009544 | |
| UN1206 | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| CCCCCCC | |
| 100.21 | |
| CHEBI:43098 | |
| ≥99% n to heptane basis (GC) |
Chemical Identifiers
| 142-82-5 | |
| 100.21 | |
| IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
| 8900 | |
| heptane |
| C7H16 | |
| MFCD00009544 | |
| n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
| CHEBI:43098 | |
| CCCCCCC |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
H225-H304-H315-H336-H410
Danger
DOTInformation : Class 3 - PG 2 - Heptanes
EINECSNumber : 205-563-8
RTECSNumber : MI7700000