missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptane, Puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC), Honeywell Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -91°C |
| Appearance | Colorless liquid |
| Density | 0.684g/mL at 25°C |
| Boiling Point | 98.2°C to 98.4°C |
| Quantity | 1 L |
| Explosion Limit | 0.07 |
| Flash Point | −4°C (24.8°F) |
| Ignition Point | 433°F |
| Packaging | Glass Bottle |
| Refractive Index | n20/D 1.3870 to 1.3880 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | P210-P261-P273-P301 + P310-P331-P501 |
| Precautionary Statement | H225-H304-H315-H336-H410 |
| DOTInformation | Class 3 - PG 2 - Heptanes |
| EINECSNumber | 205-563-8 |
| RTECSNumber | MI7700000 |