missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecylbenzene 98.0+%, TCI America™
$121.16 - $361.62
Chemical Identifiers
| CAS | 14752-75-1 |
|---|---|
| Molecular Formula | C23H40 |
| Molecular Weight (g/mol) | 316.57 |
| MDL Number | MFCD00039873 |
| InChI Key | ZMPPFNHWXMJARX-UHFFFAOYSA-N |
| Synonym | 1-Phenylheptadecane |
| PubChem CID | 84623 |
| IUPAC Name | heptadecylbenzene |
| SMILES | CCCCCCCCCCCCCCCCCC1=CC=CC=C1 |
Chemical Identifiers
| 14752-75-1 | |
| 316.57 | |
| ZMPPFNHWXMJARX-UHFFFAOYSA-N | |
| 84623 | |
| CCCCCCCCCCCCCCCCCC1=CC=CC=C1 |
| C23H40 | |
| MFCD00039873 | |
| 1-Phenylheptadecane | |
| heptadecylbenzene |
Specifications
| 14752-75-1 | |
| C23H40 | |
| 5 g | |
| ZMPPFNHWXMJARX-UHFFFAOYSA-N | |
| heptadecylbenzene | |
| 84623 | |
| ≥98.0% (GC) | |
| Heptadecylbenzene |
| White | |
| MFCD00039873 | |
| 1-Phenylheptadecane | |
| CCCCCCCCCCCCCCCCCC1=CC=CC=C1 | |
| 316.57 | |
| 316.57 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes