missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecylbenzene 98.0+%, TCI America™
Supplier: TCI America H002025G
Specifications
| Heptadecylbenzene | |
| White | |
| MFCD00039873 | |
| 1-Phenylheptadecane | |
| CCCCCCCCCCCCCCCCCC1=CC=CC=C1 | |
| 316.57 | |
| 316.57 | |
| Crystalline Lumps |
| 14752-75-1 | |
| C23H40 | |
| 25 g | |
| ZMPPFNHWXMJARX-UHFFFAOYSA-N | |
| heptadecylbenzene | |
| 84623 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 14752-75-1 | |
| 316.57 | |
| ZMPPFNHWXMJARX-UHFFFAOYSA-N | |
| 84623 | |
| CCCCCCCCCCCCCCCCCC1=CC=CC=C1 |
| C23H40 | |
| MFCD00039873 | |
| 1-Phenylheptadecane | |
| heptadecylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes