accessibility menu, dialog, popup

UserName

H3C

CAS RN 14752-75-1

IUPAC Name: heptadecylbenzene
Synonyms: benzene, heptadecyl-
Molecular Weight (g/mol): 316.57
Molecular Formula: C23H40
InChi Key: ZMPPFNHWXMJARX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results