missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptacosafluorotributylamine 60.0+%, TCI America™
Supplier: TCI America P0074100G
Specifications
| Heptacosafluorotributylamine [for Mass spectrometry] | |
| Colorless | |
| C12F27N | |
| 100 g | |
| RVZRBWKZFJCCIB-UHFFFAOYSA-N | |
| 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine | |
| 9397 | |
| 671.10 | |
| Liquid |
| 311-89-7 | |
| 178°C | |
| MFCD00000436 | |
| perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine | |
| C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F | |
| 671.096 | |
| CHEBI:38854 | |
| ≥60.0% (GC) |
Chemical Identifiers
| 311-89-7 | |
| 671.096 | |
| RVZRBWKZFJCCIB-UHFFFAOYSA-N | |
| 9397 | |
| 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine |
| C12F27N | |
| MFCD00000436 | |
| perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine | |
| CHEBI:38854 | |
| C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F |
Safety and Handling
EINECSNumber : (2)-0173
RTECSNumber : YA1000000
TSCA : Yes