missing translation for 'onlineSavingsMsg'
Learn More

Heptacosafluorotributylamine 60.0+%, TCI America™

Catalog No. p-7134409
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P007425G 25 g
P0074100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P007425G Supplier TCI America Supplier No. P007425G
Only null left

Chemical Identifiers

CAS 311-89-7
Molecular Formula C12F27N
Molecular Weight (g/mol) 671.096
MDL Number MFCD00000436
InChI Key RVZRBWKZFJCCIB-UHFFFAOYSA-N
Synonym perfluorotributylamine, tris perfluorobutyl amine, tri perfluorobutyl amine, tris nonafluorobutyl amine, heptacosafluorotributylamine, fluorinert, fluosol 43, perfluorotri-n-butylamine, fluorocarbon fc 43, tri nonafluorobutyl amine
PubChem CID 9397
ChEBI CHEBI:38854
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluoro-N,N-bis(1,1,2,2,3,3,4,4,4-nonafluorobutyl)butan-1-amine
SMILES C(C(C(F)(F)F)(F)F)(C(N(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 178°C
Quantity 25 g
Formula Weight 671.10
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptacosafluorotributylamine [for Mass spectrometry]
EINECSNumber (2)-0173
RTECSNumber YA1000000
TSCA Yes