accessibility menu, dialog, popup

UserName

CAS RN 311-89-7

IUPAC Name: tris(1,1,2,2,3,3,4,4,4-nonafluorobutyl)amine
Synonyms: perfluorotributylamine FTBA heptacosafluorotributylamine
Molecular Weight (g/mol): 671.1
Molecular Formula: C12F27N
InChi Key: RVZRBWKZFJCCIB-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)