missing translation for 'onlineSavingsMsg'
Learn More

Heptachlor, Restek

Catalog No. 06719857
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719857 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719857 Supplier Chromatographic Supplier No. RK 32228

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76-44-8
Molecular Formula C10H5Cl7
Molecular Weight (g/mol) 373.3
InChI Key FRCCEHPWNOQAEU-UHFFFAOYSA-N
PubChem CID 3589
ChEBI CHEBI:34785
SMILES C1=CC(C2C1C3(C(=C(C2(C3(Cl)Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Quantity 1 mL
Chemical Name or Material Heptachlor