accessibility menu, dialog, popup

UserName

ClClClClClClCl

CAS RN 76-44-8

IUPAC Name: 1,5,7,8,9,10,10-heptachlorotricyclo[5.2.1.0²,⁶]deca-3,8-diene
Synonyms: heptachlor heptachlor isomer 3-chlorochlordene heptachlorane heptamul
Molecular Weight (g/mol): 373.3
Molecular Formula: C10H5Cl7
InChi Key: FRCCEHPWNOQAEU-UHFFFAOYNA-N
SMILES: ClC1C=CC2C1C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results