missing translation for 'onlineSavingsMsg'
Learn More

HEPES Sodium Salt (White Powder), Fisher BioReagents

Catalog No. BP410500
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP410500 500 g Poly Bottle
BP4101 1 kg Poly Bottle
BP410212 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP410500 Supplier Fisher BioReagents Supplier No. FLBP410 500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HEPES Sodium Salt is used as a buffer for in vitro cell culture.

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance 0.04 max. (0.1M in water, 1cm) at 260nm, 0.02 max. (0.1M in water, 1cm) at 280nm
Color White
Physical Form Powder or Solid
Quantity 500 g
Assay Percent Range ≥99 %
Identification Pass Test
Loss on Drying 3.0% max.
Packaging Poly Bottle
Formula Weight 260.29
Percent Purity ≥99%
Chemical Name or Material HEPES Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Recommended Storage RT