missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HEPES, Sodium Salt, ULTROL™ Grade, Calbiochem™,

Catalog No. 391333500GM
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
391333500GM 500 g
391333100GM 100 g
3913331KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 391333500GM Supplier MilliporeSigma™ Supplier No. 391333500GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A zwitterionic N-substituted aminosulfonic acid buffer

Hygroscopic

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid, Na
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Physical Form Solid
Quantity 500 g
Solution Type Buffer
Assay Percent Range ≥99.0%
Solubility Information Soluble in water
Formula Weight 260.3
Percent Purity 99% (titration)
Grade by titration (dry basis)
Chemical Name or Material zwitterionic N-substituted aminosulfonic acid buffer
Recommended Storage 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.