missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Sodium Salt, Cell Culture Reagent Grade, ≥98.0%, MP Biomedicals™
Specifications
Specifications
| Color | White |
| pH | 9.5 - 10.8(1% aq soln) |
| Physical Form | Powder |
| Quantity | 10 g |
| Solution Type | Buffer |
| Assay Percent Range | ≥98.0% |
| Formula Weight | 260.28 |
| Percent Purity | ≥98% |
| Grade | Cell Culture |
| Chemical Name or Material | HEPES Sodium Salt |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15 to 30°C). |