missing translation for 'onlineSavingsMsg'
Learn More

HEPES Sodium Salt, Biological Buffer, High Purity, 98%, Spectrum™ Chemical

Catalog No. p-7165646
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610767 25 g Amber Glass Bottle
18610768 100 g Amber Glass Bottle
18610769 250 g Amber Glass Bottle
18610770 500 g Amber Glass Bottle
18610771 1 kg Amber Glass Bottle
18610772 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610767 Supplier Spectrum Chemical Mfg Cor Supplier No. H108425GM
Only null left

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance 0.05 A.U. (0.1 M Solution at 260 nm)
CAS Max % 1
pH Alkaline
Physical Form Crystalline
Quantity 25 g
Loss on Drying 0.04
Packaging Amber Glass Bottle
Percent Purity 98%
Grade High Purity
Chemical Name or Material 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid Sodium Salt

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.