missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Sodium Salt, Biological Buffer, High Purity, 98%, Spectrum™ Chemical
Specifications
Specifications
| Absorbance | 0.05 A.U. (0.1 M Solution at 260 nm) |
| CAS Max % | 1 |
| pH | Alkaline |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Loss on Drying | 0.04 |
| Packaging | Amber Glass Bottle |
| Percent Purity | 98% |
| Grade | High Purity |
| Chemical Name or Material | 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid Sodium Salt |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.