missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99+%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. AC327265000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327265000 500 g Plastic bottle
AC327261000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327265000 Supplier Thermo Scientific Chemicals Supplier No. 327265000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance (0.1 M in H2O), (0.1 M in H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
Absorbance Conditions (0.1M in H2O)
Melting Point 248.0°C
Color White
pH 5.0 to 15.0 (1% soln.)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Packaging Plastic bottle
Formula Weight 260.27
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material HEPES sodium salt, 99+%
Show More Show Less

RUO – Research Use Only