missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES sodium salt, 99%

Catalog No. AC215005000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC215005000 500 g
AC215000250 25 g
AC215001000 100 g
AC215000010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC215005000 Supplier Thermo Scientific Chemicals Supplier No. 215005000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 75277-39-3
Molecular Formula C8H17N2NaO4S
Molecular Weight (g/mol) 260.28
MDL Number MFCD00036463
InChI Key RDZTWEVXRGYCFV-UHFFFAOYSA-M
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt
PubChem CID 2724248
ChEBI CHEBI:46758
IUPAC Name sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
SMILES [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1

Specifications

Absorbance (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm
Absorbance Conditions (0.1M aq. soln.)
Color White
pH 9.5 to 10.5 (1% aq. soln.)
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Formula Weight 260.27
Percent Purity ≥98.5% (on anhydrous substance)
Chemical Name or Material HEPES sodium salt, 99%
Show More Show Less
EINECSNumber 278-169-7
TSCA TSCA

RUO – Research Use Only