missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
| Absorbance | (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm |
| Absorbance Conditions | (0.1M aq. soln.) |
| Color | White |
| pH | 9.5 to 10.5 (1% aq. soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Formula Weight | 260.27 |
| Show More |
Safety and Handling
| EINECSNumber | 278-169-7 |
| TSCA | TSCA |
RUO – Research Use Only