missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ HEPES, Powder
An alternative to bicarbonate for buffering
$882.76 - $882.76
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- pH 7.2 to 7.6
- Molecular Weight: 238.3
Specifications
| Powder | |
| Plastic Bottle | |
| Sterile |