Learn More
Thermo Scientific Chemicals HEPES hemisodium salt
A zwitterionic buffer | CAS: 103404-87-1 | C8H17N2NaO4S | 260.28 g/mol
Supplier: Thermo Scientific Chemicals J6183036
Description
HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Zwitterionic Buffer | |
| White | |
| 500 g | |
| C8H17N2NaO4S | |
| 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
| 2724248 | |
| 497.58 |
| 103404-87-1 | |
| Crystalline Powder | |
| 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic Acid Sodium Salt | |
| MFCD00036463 | |
| Soluble in water. | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
| 260.28 | |
| CHEBI:46758 |
Chemical Identifiers
| 103404-87-1 | |
| 260.28 | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| 2724248 | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| C8H17N2NaO4S | |
| MFCD00036463 | |
| 4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt | |
| CHEBI:46758 | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only