missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES hemisodium salt

A zwitterionic buffer | CAS: 103404-87-1 | C8H17N2NaO4S | 260.28 g/mol

Supplier:  Thermo Scientific Chemicals J6183030

 View more versions of this product

Catalog No. AAJ6183030


Only null left

Description

Description

HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES hemisodium salt is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

HEPES hemisodium salt
White
250 g
MFCD00036463
Soluble in water.
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
260.28
CHEBI:46758
103404-87-1
Crystalline Powder
C8H17N2NaO4S
4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt
RDZTWEVXRGYCFV-UHFFFAOYSA-M
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
2724248
497.58

Chemical Identifiers

103404-87-1
260.28
RDZTWEVXRGYCFV-UHFFFAOYSA-M
2724248
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
C8H17N2NaO4S
MFCD00036463
4-(2-Hydroxyethyl)-1-piperazineethanesulfonic acid sodium salt
CHEBI:46758
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only