missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES-buffered saline (2X, pH 7.0), low endotoxin
$143.52 - $143.52
Specifications
| Chemical Name or Material | HEPES-buffered saline (2X, pH 7.0) |
|---|---|
| Name Note | low endotoxin |
| Color | Colorless |
| Physical Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ67505AE
|
Thermo Scientific Chemicals
J67505AE |
100 mL |
Each for $143.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| HEPES-buffered saline (2X, pH 7.0) | |
| low endotoxin | |
| Colorless | |
| Liquid |
RUO – Research Use Only