missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.
Specifications
Specifications
| Chemical Name or Material | HEPES Buffer |
| Absorbance | ≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm |
| Name Note | 1M Solution, pH 7.3 |
| CAS | 7365-45-9, 7732-18-5 |
| Color | Undesignated |
| Concentration | 0.95 to 1.05 M |
| pH | 7.2 to 7.5 |
| Physical Form | Liquid |
| Quantity | 100 mL |
| DNase | DNase free |
| Show More |