missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Hematoxylin, 85%, pure, certified
CAS: 517-28-2 | C16H14O6 | 302.28 g/mol
$643.27
Chemical Identifiers
| CAS | 517-28-2 |
|---|---|
| Molecular Formula | C16H14O6 |
| Molecular Weight (g/mol) | 302.28 |
| MDL Number | MFCD00078111 |
| InChI Key | WZUVPPKBWHMQCE-WKTCHCBJNA-N |
| Synonym | Natural Black 1, C.I. 75290 |
| PubChem CID | 45029742 |
| IUPAC Name | (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol |
| SMILES | [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC411161000
|
Thermo Scientific Chemicals
411161000 |
100 g | Glass bottle |
Each for $643.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 517-28-2 | |
| 302.28 | |
| WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
| 45029742 | |
| [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 |
| C16H14O6 | |
| MFCD00078111 | |
| Natural Black 1, C.I. 75290 | |
| (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol |
Specifications
| 100.0°C to 120.0°C | |
| 85% | |
| MFCD00078111 | |
| Solubility in water: slightly soluble; soluble in hot water;. Other solubilities: soluble in hot alcohol, glycerol, borax, alkali hy, droxides (aq. solution purple red; becomes blue vi, olett during storage at air). | |
| [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3 | |
| (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol | |
| 45029742 | |
| 10% (105°C) max. | |
| Pure | |
| Beige to Brown | |
| Hematoxylin |
| 517-28-2 | |
| C16H14O6 | |
| Natural Black 1, C.I. 75290 | |
| WZUVPPKBWHMQCE-WKTCHCBJNA-N | |
| Authentic | |
| 302.28 | |
| 302.27 | |
| 85% | |
| 88% (on dry substance) min. | |
| Powder |
Safety and Handling
RUO – Research Use Only