missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Hematoxylin, 85%, pure, certified

Catalog No. AC411161000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411161000 100 g Glass bottle
AC411165000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411161000 Supplier Thermo Scientific Chemicals Supplier No. 411161000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black 1, C.I. 75290
PubChem CID 45029742
IUPAC Name (6aR)-7,11b-dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

Melting Point 100.0°C to 120.0°C
Assay Percent Range 85%
Solubility Information Solubility in water: slightly soluble; soluble in hot water;. Other solubilities: soluble in hot alcohol, glycerol, borax, alkali hy, droxides (aq. solution purple red; becomes blue vi, olett during storage at air).
Infrared Spectrum Authentic
Formula Weight 302.27
Loss on Drying 10% (105°C) max.
Percent Purity 85%
Grade Pure
Packaging Glass bottle
UV_Vis 88% (on dry substance) min.
Color Beige to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Hematoxylin
Show More Show Less
Hazard Category

RUO – Research Use Only