missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Heavy Designed Polystyrene Vented Petri Dishes is designed for better heat resistance using hot agar.
Supplier: Greiner Bio-One 664102
Description
- Vented for better gas exchange
- Heavy design for heat resistance using hot agar
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
| Clear | |
| Round | |
| Petri Dish | |
| Yes | |
| 100 mm |
| Polystyrene | |
| Non-sterile | |
| 360 | |
| 20 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.