missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Glass Headspace Vial with Beveled Top
Available in several sizes, the style and diameter of headspace vials depend on the autosampler.
Supplier: MilliporeSigma™ Supelco™ 27296
Description
Chemical Identifiers
| Clear | |
| Glass | |
| Rounded Bottom |
| 22.6 mm | |
| Beveled |
Specifications
| Clear | |
| Glass | |
| Beveled | |
| Rounded Bottom |
| 22.6 mm | |
| 20 mL | |
| 100 | |
| 75 mm |