missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HCT 116 (+hIGF-1) Control Lysate (1mg/mL)

Catalog No. PIA37961
Change view
Click to view available options
Quantity:
0.5 mg (1 mg/mL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA37961 0.5 mg (1 mg/mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA37961 Supplier Thermo Scientific™ Supplier No. A37961

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ HCT 116 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the RAS signaling pathway.

  • Validated for the expression RAS by western blotting and of NRAS, KRAS, and HRAS by mass spectrometry analysis
  • Optimized - stimulation and lysis conditions optimized for detection of RAS signaling targets
  • Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
  • Stable for two years at -80°C

Specifications

Detection Method Mass Spectrometry
For Use With (Equipment) Mass Spectrometer
Shipping Condition Dry Ice
Workflow Step Sample Enrichment
Form Frozen Liquid
Product Line Thermo Scientific
Quantity 0.5 mg (1 mg/mL)
Type Control Lysate
Final Product Type Protein
Starting Material Cell Lysate
What are the characteristics of HCT 116 (+hIGF-1) Control Lysate, and what is its main purpose?

HCT 116 (+hIGF-1) Control Lysate is a stimulated HCT 116 cell lysate that has been validated for the expression levels and phosphorylation of key targets (NRAS, KRAS, and HRAS) in the RAS signaling pathway. It serves as a positive control for use with SureQuant RAS panel kits.

For Research Use Only. Not for use in diagnostic procedures.