missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corepoint™ Scientific Hazardous Location Manual Defrost Freezer
Delivering professional protection for cold storage in hazardous laboratory locations.
$9,607.94 - $10,448.33
Specifications
| Type | Explosion Proof Freezer |
|---|---|
| Temperature Range | -15°C to -25°C |
| Door Style | Solid Swing |
| No. of Doors | 1 |
| Cabinet Material | Scratch-resistant Painted Steel |
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Price | Quantity & Availability | |||||
|
361058663
|
Corepoint™ Scientific
EF141WWW/0MHC |
14 cu. ft. |
Each for $9,644.61
|
|
|||||
|
361058665
|
Corepoint™ Scientific
EF171WWW/0MHC |
17 cu. ft. |
Each for $9,607.94
|
|
|||||
|
361058668
|
Corepoint™ Scientific
EF201WWW/0MHC |
20 cu. ft. |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Hazardous location freezers meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines.
- These units utilize nonsparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
- Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
- Our freezers run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
- For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see Flammable Material Storage units.
Specifications
| Explosion Proof Freezer | |
| Solid Swing | |
| Scratch-resistant Painted Steel | |
| Cabinet foamed-in-place with EPA compliant high density urethane foam | |
| White Powder Coated Steel | |
| cULus, ETL, ENERGYSTAR | |
| White | |
| Two (2) Year Parts and Labor Warranty | |
| Microprocessor Temperature Controller | |
| 115 V | |
| Natural Hydrocarbon Refrigerant |
| -15°C to -25°C | |
| 1 | |
| Painted Steel | |
| Painted Steel | |
| 6 | |
| Upright | |
| Fan-forced | |
| Manual | |
| No Plug, Hardwired | |
| H-drive |