missing translation for 'onlineSavingsMsg'
Learn More

Corepoint™ Scientific Hazardous Location Manual Defrost Freezer

Delivering professional protection for cold storage in hazardous laboratory locations.

Supplier:  Corepoint™ Scientific EF171WWW/0MHC

 View more versions of this product

Catalog No. 361058665


Only null left

Description

Description

Hazardous location freezers meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines.

  • These units utilize nonsparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
  • Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
  • Our freezers run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
  • For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see Flammable Material Storage units.
Specifications

Chemical Identifiers

Explosion Proof Freezer
Solid Swing
Scratch-resistant Painted Steel
Cabinet foamed-in-place with EPA compliant high density urethane foam
White Powder Coated Steel
cULus, ETL, ENERGYSTAR
White
Two (2) Year Parts and Labor Warranty
Microprocessor Temperature Controller
115 V
Natural Hydrocarbon Refrigerant
-15°C to -25°C
1
Painted Steel
Painted Steel
6
Upright
Fan-forced
Manual
No Plug, Hardwired
H-drive

Specifications

Explosion Proof Freezer
481 L
Solid Swing
Scratch-resistant Painted Steel
Cabinet foamed-in-place with EPA compliant high density urethane foam
White Powder Coated Steel
cULus, ETL, ENERGYSTAR
White
Two (2) Year Parts and Labor Warranty
EF171WWW/0MHC
5.5 Amps
115 V
Natural Hydrocarbon Refrigerant
17 cu. ft.
-15°C to -25°C
1
Painted Steel
Painted Steel
6
Upright
Fan-forced
Manual
Microprocessor Temperature Controller
No Plug, Hardwired
H-drive
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions