Learn More
New Pig Haz-Mat Socks
Stop the spread of acids and bases.
Supplier: New Pig PIG301
Description
- Chemical-resistant — won't degrade or cause a dangerous reaction upon contact with corrosive spills
- Excellent for quickly diking and stopping the spread of chemical spills
- Absorb and retain most acids, bases and unknown liquids
- Can be incinerated after use to reduce waste
Specifications
| Pink | |
| 7.6 L | |
| Hazmat Absorbent Sock | |
| 10 ft. L x 3 in. W | |
| 10 ft. | |
| 3 in. | |
| 3 in. | |
| 14 lb. |
| 2 gal. | |
| Polypropylene | |
| Quickly diking and stopping the spread of chemical spills, absorbs acids, bases, corrosives | |
| 10 ft. x 3 in. (3.04 m x 7.62 cm) | |
| 3.04 m | |
| 7.62 cm | |
| 7.62 cm | |
| 6.4 kg |