missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Presterilized

Black tips for accurate sample delivery

$963.61 - $4233.30

Specifications

Sterility Sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
View More Specs

Products 8
Catalog Number Mfr. No. Volume (Metric) Filtered Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Filtered Price Quantity  
14-222-859
View Documents
Axygen™
TTF50CBKHTRS36
25 μL Filtered
Case of 6912 Each for $4,233.30
Only null left
 
14-222-853
View Documents
Axygen™
TTF200CBKHTRS
200 μL Filtered
Case of 2304 Each for $1,708.96
Only null left
 
14-222-856
View Documents
Axygen™
TTF20CBKHTRS
20 μL Filtered
Case of 2304 Each for $1,720.83
Only null left
 
14-222-847
View Documents
Axygen™
TT50CBKHTRS
50 μL Non-filtered
Case of 2304 Each for $1,487.78
Only null left
 
14-222-837
View Documents
Axygen™
TT200CBKHTRS
200 μL Non-filtered
Case of 2304 Each for $1,488.13
Only null left
 
14-222-851
View Documents
Axygen™
TTF1000CBKHTRS
1000 μL Filtered
Case of 1536 Each for $1,033.17
Only null left
 
14-222-858
View Documents
Axygen™
TTF50CBKHTRS
50 μL Filtered
Case of 2304 Each for $1,446.93
Only null left
 
14-222-832
View Documents
Axygen™
TT1000CBKHTRS
1000 μL Non-filtered
Case of 1536 Each for $963.61
Only null left
 
Description

Description

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Liquid Level Sensing Tips
  • 96 Tip format
  • Conductive Black Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head, PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Specifications

    Sterile
    Autoclavable
    For accurate sample delivery
    Automation, Round
    Polypropylene
    Black
    Non-ANSI
    Rack
    Hanging Tip Rack
    Videos
    SDS
    missing translation for 'documents'

    missing translation for 'documents'