missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Presterilized
GREENER_CHOICE

Catalog No. 14222851
Change view
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
200 μL
1000 μL
Filtered:
Filtered
Non-filtered
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered
14222851 1000 μL Filtered
14222859 25 μL Filtered
14222853 200 μL Filtered
14222856 20 μL Filtered
14222847 50 μL Non-filtered
14222837 200 μL Non-filtered
14222858 50 μL Filtered
14222832 1000 μL Non-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14222851 Supplier Axygen™ Supplier No. TTF1000CBKHTRS
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Black tips for accurate sample delivery

Axygen™ offers a comprehensive line of tips for Tecan™, Aurora Biomed and PerkinElmer™ automation systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Tips are RNase-/DNase-free and nonpyrogenic. Available in nonsterile, sterile, non-filtered and filtered. Clear and Conductive black polypropylene tips are available in 96-well format and is also available in hanging tip rack format designed for Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head and PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head.


  • 1000 μL Liquid Level Sensing Tips
  • 96 Tip format
  • Conductive Black Polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Sterile
  • Compatibility: Tecan™ Genesis Freedom™/Freedom Eva™/Miniprep with LiHa-1, -4, -8 head, Aurora Biomed Versa™ with 8-Channel head, PerkinElmer™ Janus™/MultiProbe with Variospan-1, 4, 8 head
  • Certifications

    Certified DNAse-, RNAse- and Pyrogen-safe

    Specifications

    Volume (Metric) 1000 μL
    Filtered Filtered
    Sterility Sterile
    Color Black
    Autoclavable Autoclavable
    Certifications/Compliance Non-ANSI
    For Use With (Application) For accurate sample delivery
    Format Rack
    Tip Style Automation, Round
    Packaging Hanging Tip Rack
    Quantity 16 Racks/Cs.
    For Use With (Equipment) Aurora Biomed Versa 8-channel Head, PerkinElmer MultiProbe Variospan-1, 4, 8 Head
    Material Polypropylene
    Show More Show Less