missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile

Black tips for accurate sample delivery

$850.57 - $3479.96

Specifications

Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
View More Specs

Products 6
Catalog Number Mfr. No. Volume (Metric) Format Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Format Price Quantity  
14-222-830
View Documents
Axygen™
TT1000CBKHTR
1000 μL Hanging Tip Racked
Case of 1536 Each for $850.57
Only null left
 
14-222-831
View Documents
Axygen™
TT1000CBKHTR24
1000 μL Hanging Tip Racked, Cardboard
Case of 4608 Each for $2,717.52
Only null left
 
14-222-835
View Documents
Axygen™
TT200CBKHTR
200 μL Hanging Tip Racked
Case of 2304 Each for $1,371.74
Only null left
 
14-222-840
View Documents
Axygen™
TT20CBKHTR
20 μL Hanging Tip Racked
Case of 2304 Each for $1,340.43
Only null left
 
14-222-845
View Documents
Axygen™
TT50CBKHTR
50 μL Hanging Tip Racked
Case of 2304 Each for $1,340.43
Only null left
 
14-222-846
View Documents
Axygen™
TT50CBKHTR36
50 μL Hanging Tip Racked, Cardboard
Case of 6912 Each for $3,479.96
Only null left
 
Description

Description

  • Polypropylene
  • Volume ranging from 20 to 1000μL
  • Available with or without aerosol-resistant filters
  • Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems

Certifications

Certified DNAse-, RNAse- and Pyrogen-safe

Specifications

Specifications

Non-sterile
Autoclavable
For accurate sample delivery
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
Black
Non-ANSI
Automation, Round
Polypropylene
Videos
SDS
Documents

Documents