Learn More
Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
Black tips for accurate sample delivery
Supplier:  Axygen™ TT50CBKHTR
 View more versions of this product
Catalog No. 14-222-845
Only null left
Explore available promotions
Description
Description
  • Polypropylene
  • Volume ranging from 20 to 1000μL
  • Available with or without aerosol-resistant filters
  • Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems
Specifications
Specifications
50 μL
Non-sterile
Autoclavable
For accurate sample delivery
Automation, Round
Polypropylene
Non-filtered
Black
Non-ANSI
Hanging Tip Racked
Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
Product Suggestions
Product Suggestions