missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Hanging Wafer Robotic Tips: Tecan-Style Liquid Level Sensing Tips, Nonsterile
GREENER_CHOICE

Catalog No. 14222830
Change view
Click to view available options
Volume (Metric):
20 μL
50 μL
200 μL
1000 μL
Format:
Hanging Tip Racked
Hanging Tip Racked, Cardboard
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format
14222830 1000 μL Hanging Tip Racked
14222831 1000 μL Hanging Tip Racked, Cardboard
14222835 200 μL Hanging Tip Racked
14222840 20 μL Hanging Tip Racked
14222845 50 μL Hanging Tip Racked
14222846 50 μL Hanging Tip Racked, Cardboard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14222830 Supplier Axygen™ Supplier No. TT1000CBKHTR
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Black tips for accurate sample delivery

  • Polypropylene
  • Volume ranging from 20 to 1000μL
  • Available with or without aerosol-resistant filters
  • Compatible with Axygen™ Tecan™/Perkin Elmer/Auroroa™ Biomed/Corbett Cas-1200 systems

Certifications

Certified DNAse-, RNAse- and Pyrogen-safe

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
For Use With (Application) For accurate sample delivery
Format Hanging Tip Racked
Tip Style Automation, Round
For Use With (Equipment) Tecan Genesis, Tecan Freedom, Tecan Freedom EVO, Tecan Miniprep LiHa-1, 4, 8 Head, Aurora Biomed Versa 8-channel Head
Material Polypropylene