missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Syringe, Luer Tip (LT), Sold by MilliporeSigma™ Supelco™
Tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip. Needles are not included.
Supplier: Merck Emd Millipore 20702
Specifications
| Autoclavable | |
| 0.061 in. | |
| 1.46 mm | |
| Microliter™ Syringe | |
| 239°F | |
| 50°F | |
| 10°C | |
| 100 μL | |
| Hamilton 710 LT |
| Borosilicate Glass (Barrel) | |
| 0.305 in. | |
| 7.75 mm | |
| 2000 psig (137.9 bar) | |
| 115°C | |
| 710 LT | |
| 1 Ea. | |
| Hamilton | |
| Luer Tip, Kel-F Hub |
Hamilton is a registered trademark of Hamilton Co.