missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP™-X200 Cation Exchange Columns

Catalog No. 1481616
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
1481616 250 mm
1481615 150 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1481616 Supplier Hamilton™ Supplier No. HAM79442

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analyze inorganic and organic cations using conductivity or UV detection. Separate mono or divalent cations depending on mobile phase conditions from 20 ppb to 200 ppm.

Can be used with any existing HPLC or ion chromatograph to easily determine cations in air, water and soil. PRP-X200 columns provide a number of benefits

Better than Wet Chemical Methods

  • Ion chromatography will greatly reduce sample pretreatment and improve the accuracy and precision of results.
  • High Sensitivity
    • Separate ions at concentrations from 20ppb to 20ppm without suppression or elaborate preconcentration techniques
  • Long Column Life
    • Highly inert polymeric support resists chemical attack from organic solvents and aqueous buffers (0-100% aqueous or organic; pH 1 to 13), effectively lengthening column life. If column performance should deteriorate (peak broadening and a loss of symmetry), a regeneration protocol will usually return your chromatography to its original state.
  • Analyze Mono or Divalent Cations
    • Quickly and easily separate five monovalent cations in five minutes or four divalent cations in four minutes
  • Unique Selectivity
    • Highly inert poly(styrene-divinylbenzene) packing allows the use of organic modifier. By varying the concentration of methanol the resolution between cations can be altered.

    • One particle size: 10°m
    • Four column internal diameters: 2.1 to 10.0mm
    • Two column materials: 316 stainless steel and PEEK
    • Analytical and semiprep/preparative guard columns

    Specifications

    Column Format Analytical Column
    Diameter (Metric) 4.1 mm
    Flow Rate 2 mL/min
    For Use With Analyzing Inorganic and Organic Cations Using Conductivity or UV Detection
    Length (Metric) 250 mm
    Max. Pressure 5000 psi (345 bar)
    Mobile Phase Compatibility pH 1 to 13, 0 to 100% aqueous, organic modifier
    Particle Size 10 μm
    pH 1 to 13
    Pore Size 100 Å
    Temperature 5°C to 60°C
    USP Type L22
    Stationary Phase PRP-X200
    Product Line PRP-X200
    Packing Material PSDVB/Sulfonic Acid
    Column Type Ion Exchange
    Phase Ion Exchange
    Material Stainless Steel
    Technique Cation Exchange
    Product Type Column
    Show More Show Less