missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Hamilton™ PRP-1 HPLC Guard Cartridge Kit, 10 μ Particle Size

Catalog No. 111009530
Change view
Click to view available options
D: 2.1 mm; L: 20 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111009530 D: 2.1 mm; L: 20 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009530 Supplier MilliporeSigma™ Supelco™ Supplier No. 82033U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit includes 1 holder and 2 cartridges

Specifications

Quantity 1 Kit
Particle Size 10 μm
Length (Metric) 20 mm
Pore Size 100 Å
Type Guard Cartridge Kit
Diameter (Metric) Inner 2.1 mm
Includes 1 holder and 2 cartridges
Material Stainless Steel Hardware
For Use With (Application) HPLC
Column Type HPLC Guard Cartridge
Product Line Hamilton
Show More Show Less