Learn More
Hamilton™ Preflanged PTFE Tubing Assemblies
Pre-made flanged tubing assemblies made with PTFE tubing and Kel-F threaded fittings. The flanged tubing assemblies provide the perfect seal with 1/4-28 flat bottom valve ports.
$103.26 - $120.47
Specifications
Material PTFE
Includes 0.25 to 28 flat bottom valve ports
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Overall Length (Metric) Gauge Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Overall Length (Metric) Gauge Price Quantity & Availability  
361012603
View Documents Promotion Details
Hamilton™
88966
Tubing Assembly; 1M6X1 FTG TPRD TIP 650 mm 18 ga
Each for $111.26
Only null left
14819163
View Documents Promotion Details
Hamilton™
HAM88942
Tubing Assembly; 1/4-28 NT (SR) 762 mm 12 ga
Each for $103.26
Only null left
14819164
View Documents Promotion Details
Hamilton™
HAM88939
Tubing Assembly; 1/4-28 NT (SR) 762 mm 18 ga
Each for $120.47
Only null left
Description
Description
Specifications
Specifications
PTFE
0.25 to 28 flat bottom valve ports