Learn More
Hamilton™ Preflanged PTFE Tubing Assemblies
Pre-made flanged tubing assemblies made with PTFE tubing and Kel-F threaded fittings. The flanged tubing assemblies provide the perfect seal with 1/4-28 flat bottom valve ports.
Supplier:  Hamilton™ HAM88942
 View more versions of this product
Catalog No. 14819163
Only null left
Description
Description
Specifications
Chemical Identifiers
PTFE
0.25 to 28 flat bottom valve ports
Specifications
PTFE
0.25 to 28 flat bottom valve ports
Tubing Assembly; 1/4-28 NT (SR)
762 mm
12 ga
Product Suggestions
Product Suggestions