Learn More
Hamilton™ Metal Hub Side Port Needles (Luer Lock)
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. Needles are 304 stainless steel (sst.). Standard needle length is 2 in. (51mm). Custom needle lengths and point styles are available.
Supplier:  Hamilton™ HAM772903
 View more versions of this product
Catalog No. 14815491
Only null left
Description
Description
Metal Hub Compatible Syringe Terminations
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
  • Needle Point Styles
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
    /li>
    Six needles per pack.
    Specifications
    Chemical Identifiers
    Autoclavable
    2 in.
    Cone with Side Port
    Nickel-plated Brass (Hub), Stainless Steel
    Replacement Needle
    TLL Syringes and LT or TLL Connectors
    Specifications
    Autoclavable
    26 G
    0.02 in.
    0.46 mm
    Replacement Needle
    TLL Syringes and LT or TLL Connectors
    Nickel-plated Brass (Hub), Stainless Steel
    0.01 in.
    0.26 mm
    2 in.
    Cone with Side Port
    Product Suggestions
    Product Suggestions