Learn More
Hamilton™ Metal Hub Blunt Point Needles (Luer Lock)
Can be used with TLL syringes and LT or TLL connectors.
Supplier:  Hamilton™ HAM91039
 View more versions of this product
Catalog No. 14-815-623
Only null left
Explore available promotions
Description
Description
Can be used with TLL syringes and LT or TLL connectors.
  • Made of 304 Stainless Steel
  • Standard needle length is 51mm (2 in.)
  • Available in convenient six packs
  • Custom needle lengths and point styles are available
  • Needles are available from 10 to 33 ga
Specifications
Specifications
Autoclavable
26s G
0.018 in.
0.45 mm
Needle
Blunt
Nickel-plated Brass (Hub), Stainless Steel
0.005 in.
0.13 mm
2 in.
51 mm
PTFE Luer-lock Syringes and Luer-tip or PTFE Luer-lock Connectors
Product Suggestions
Product Suggestions